Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxybenzoyl chloride ≥98.0%

Supplier: Decon Laboratories Ltd

E0700-25G E0700-5G
TCE0700-25GEA 40.65 USD InStock
TCE0700-25G TCE0700-5G
4-Ethoxybenzoyl chloride ≥98.0%
4-Ethoxybenzoyl chloride
CAS Number: 16331-46-7
MDL Number: MFCD00000688
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 145
Flash Point (°C): 110
Freezing point (°C): 22


Formula: C₉H₉ClO₂
Boiling Pt: 145 °C
Flash Pt: 110 °C
MDL Number: MFCD00000688
CAS Number: 16331-46-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=124f74923c4b65b14ec44158b2330acc3f7145cc&vid=124f74923c4b65b14ec44158b2330acc3f7145cc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty