Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoroanisole ≥99.0% (by GC)

Supplier: Decon Laboratories Ltd

F0254-10G F0254-25G
TCF0254-10GEA 25 USD InStock
TCF0254-10G TCF0254-25G
3-Fluoroanisole ≥99.0% (by GC)
3-Fluoroanisole
CAS Number: 456-49-5
MDL Number: MFCD00000335
Molecular Formula: C7H7FO
Molecular Weight: 126.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 43
Specific Gravity (20/20): 1.11


Formula: C₇H₇FO
Boiling Pt: 158 °C
Density: 1.11 (20 °C)
Flash Pt: 43 °C
MDL Number: MFCD00000335
CAS Number: 456-49-5
UN: 1993
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36cc05b3c5b0d7ea51137400ad07642132f67be7&vid=36cc05b3c5b0d7ea51137400ad07642132f67be7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty