Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥95.0%

Supplier: Decon Laboratories Ltd

F0358-25G F0358-5G
TCF0358-25GEA 105.18 USD InStock
TCF0358-25G TCF0358-5G
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥95.0%
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)
CAS Number: 140681-55-6
MDL Number: MFCD00142607
Molecular Formula: C7H14B2ClF9N2
Molecular Weight: 354.26
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 175
Flash Point (°C): 55

Product Categories: Synthetic Organic Chemistry, Fluorinating Agents
Other Product Names / Synonyms: 1-(Chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane Bis(tetrafluoroborate), F-TEDA-BF4


Formula: C₇H₁₄B₂ClF₉N₂
MW: 354.26 g/mol
Melting Pt: 175 °C
Flash Pt: 55 °C
MDL Number: MFCD00142607
CAS Number: 140681-55-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4712fb52dd86b1b6d66230bd1f83e68658b9450e&vid=4712fb52dd86b1b6d66230bd1f83e68658b9450e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty