Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluorotriphenylsilane ≥97.0%

Supplier: Decon Laboratories Ltd

F0439-25G F0439-5G
TCF0439-25GEA 285 USD InStock
TCF0439-25G TCF0439-5G
Fluorotriphenylsilane ≥97.0%
Fluorotriphenylsilane
CAS Number: 379-50-0
MDL Number: MFCD00017899
Molecular Formula: C18H15FSi
Molecular Weight: 278.40
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 64

Other Product Names / Synonyms: Triphenylfluorosilane, Triphenylsilyl Fluoride


Formula: C₁₈H₁₅FSi
MW: 278.4
Boiling Pt: 210 °C
Melting Pt: 63
Density: 1.212
MDL Number: MFCD00017899
CAS Number: 379-50-0
UN: 3261
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39f978b3cfebb431bdc31af8d55bf2e6fed19a30&vid=39f978b3cfebb431bdc31af8d55bf2e6fed19a30&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty