Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorostyrene ≥98.0% (by GC) stabilized

Supplier: Decon Laboratories Ltd

F0461-25G F0461-5G
TCF0461-25GEA 240 USD InStock
TCF0461-25G TCF0461-5G
4-Fluorostyrene ≥98.0% (by GC) stabilized
4-Fluorostyrene
(stabilized with TBC)
CAS Number: 405-99-2
MDL Number: MFCD00000361
Molecular Formula: C8H7F
Molecular Weight: 122.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): 35
Freezing point (°C): -35
Specific Gravity (20/20): 1.02
Storage Temperature: 0-10°C


Formula: C₈H₇F
Boiling Pt: 67 °C
Density: 1.02 (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00000361
CAS Number: 405-99-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=041b7710ace0e72696caac2ad652482fc0d48b56&vid=041b7710ace0e72696caac2ad652482fc0d48b56&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty