Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-4-ethynylbenzene ≥98.0%

Supplier: Decon Laboratories Ltd

F0470-1G F0470-5G
TCF0470-1GEA 47.83 USD InStock
TCF0470-1G TCF0470-5G
1-Fluoro-4-ethynylbenzene ≥98.0%
1-Fluoro-4-ethynylbenzene
CAS Number: 766-98-3
MDL Number: MFCD00168823
Molecular Formula: C8H5F
Molecular Weight: 120.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 56
Flash Point (°C): 29
Freezing point (°C): 24
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 4-Fluorophenylacetylene


Formula: C₈H₅F
MW: 120.13
Boiling Pt: 56 °C
Flash Pt: 29 °C
MDL Number: MFCD00168823
CAS Number: 766-98-3
UN: 1325
ADR: 4.1 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bc1019f5351696ec6cc889ccb9d09c095da7812&vid=5bc1019f5351696ec6cc889ccb9d09c095da7812&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty