Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorobenzylamine ≥98.0%

Supplier: Decon Laboratories Ltd

F0538-25G F0538-5G
TCF0538-25GEA 150 USD InStock
TCF0538-25G TCF0538-5G
2-Fluorobenzylamine ≥98.0%
2-Fluorobenzylamine
CAS Number: 89-99-6
Molecular Formula: C7H8FN
Molecular Weight: 125.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 75
Flash Point (°C): 67
Specific Gravity (20/20): 1.11


Formula: C₇H₈FN
Boiling Pt: 75 °C
Density: 1.11 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00008107
CAS Number: 89-99-6
UN: 2735
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3f2698a43fa8dfa52e3adb97b80be7119d02382&vid=e3f2698a43fa8dfa52e3adb97b80be7119d02382&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty