Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glyoxylic acid ∼50% ∼9 mol/L in water

Supplier: Decon Laboratories Ltd

G0366-25ML G0366-500ML
TCG0366-25MLEA 37 USD InStock
TCG0366-25ML TCG0366-500ML
Glyoxylic acid ∼50% ∼9 mol/L in water
Glyoxylic acid
CAS Number: 298-12-4
MDL Number: MFCD00006958
Molecular Formula: C2H2O3
Molecular Weight: 74.04
Form: Clear Liquid
Color: Colorless


Formula: C₂H₂O₃
MW: 74.04 g/mol
MDL Number: MFCD00006958
CAS Number: 298-12-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2fb5238013e2c9070938782ca61c4713047711a&vid=c2fb5238013e2c9070938782ca61c4713047711a&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty