Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroxyurea ≥97.0%

Supplier: Decon Laboratories Ltd

H0310-25G H0310-5G
TCH0310-25GEA 304.4 USD InStock
TCH0310-25G TCH0310-5G
Hydroxyurea ≥97.0%
Hydroxyurea
CAS Number: 127-07-1
MDL Number: MFCD00007943
Molecular Formula: CH4N2O2
Molecular Weight: 76.06
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 140


Formula: CH₄N₂O₂
MW: 76.05 g/mol
Boiling Pt: 296
Melting Pt: 140 °C
MDL Number: MFCD00007943
CAS Number: 127-07-1
Merck Index: 13,04874
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ed517e3a1c595ebb347571735e9ecf5d85cefea&vid=5ed517e3a1c595ebb347571735e9ecf5d85cefea&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty