H0476-25G
H0476-5G
TCH0476-25GEA
153
USD
InStock
TCH0476-25G
TCH0476-5G
1,1,1,5,5,5-Hexafluoroacetylacetone ≥95.0%
1,1,1,5,5,5-Hexafluoroacetylacetone
CAS Number: 1522-22-1
MDL Number: MFCD00000426
Molecular Formula: C5H2F6O2
Molecular Weight: 208.06
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): 32
Specific Gravity (20/20): 1.49
Product Categories: Synthetic Organic Chemistry, Ligands, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione