Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexamethyldisilathiane ≥97.0%

Supplier: Decon Laboratories Ltd

H0871-25G H0871-5G
TCH0871-25GEA 555.42 USD InStock
TCH0871-25G TCH0871-5G
Hexamethyldisilathiane ≥97.0%
Hexamethyldisilathiane
CAS Number: 3385-94-2
MDL Number: MFCD00014851
Molecular Formula: C6H18SSi2
Molecular Weight: 178.44
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 163
Flash Point (°C): 23
Specific Gravity (20/20): 0.85

Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: 1,1,1,3,3,3-Hexamethyldisilathiane, Thiobis(trimethylsilane)


Formula: C₆H₁₈SSi₂
Boiling Pt: 163 °C
Density: 0.85 (20 °C)
Flash Pt: 23 °C
MDL Number: MFCD00014851
CAS Number: 3385-94-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc31166238c9427eea5b66a3f5f5015b021911&vid=13cc31166238c9427eea5b66a3f5f5015b021911&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty