Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexakis(bromomethyl)benzene ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

H0901-1G H0901-5G
TCH0901-1GEA 48 USD InStock
TCH0901-1G TCH0901-5G
Hexakis(bromomethyl)benzene ≥98.0% (by HPLC, titration analysis)
Hexakis(bromomethyl)benzene
CAS Number: 3095-73-6
MDL Number: MFCD00182539
Molecular Formula: C12H12Br6
Molecular Weight: 635.65
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Formula: C₁₂H₁₂Br₆
MDL Number: MFCD00182539
CAS Number: 3095-73-6
UN: 3261
ADR: 8,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f2bb87e545a3f8962a925225659102c9f70e42a&vid=5f2bb87e545a3f8962a925225659102c9f70e42a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty