Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexadecyltrimethylammonium hydroxide 10% in water

Supplier: Decon Laboratories Ltd

H0974-25ML H0974-500ML
TCH0974-25MLEA 51.75 USD InStock
TCH0974-25ML TCH0974-500ML
Hexadecyltrimethylammonium hydroxide 10% in water
Hexadecyltrimethylammonium hydroxide
CAS Number: 505-86-2
MDL Number: MFCD00041921
Molecular Formula: C19H43NO
Molecular Weight: 301.56
Form: Clear Liquid
Color: Colorless

Other Product Names / Synonyms: Cetyltrimethylammonium Hydroxide


Formula: C₁₉H₄₃NO
MW: 301.56 g/mol
MDL Number: MFCD00041921
CAS Number: 505-86-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e93de71049dbf18dfe0a19a2f5f96c11bfc8a325&vid=e93de71049dbf18dfe0a19a2f5f96c11bfc8a325&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty