Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexyl nitrite ≥95.0%

Supplier: Decon Laboratories Ltd

H1066-25ML H1066-500ML
TCH1066-25MLEA 63.69 USD InStock
TCH1066-25ML TCH1066-500ML
Hexyl nitrite ≥95.0%
Hexyl nitrite
CAS Number: 638-51-7 MDL Number: MFCD06797110 Molecular Formula: C6H13NO2 Molecular Weight: 131.18 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 52 Specific Gravity (20/20): 0.88 Storage Temperature: 0-10°C


Formula: C₆H₁₃NO₂
MW: 131.18 g/mol
Boiling Pt: 52 °C
Density: 0.88 (20 °C)
MDL Number: MFCD06797110
CAS Number: 638-51-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc7c42ca7357572f509cfd8d6298aa8e7a6955b4&vid=fc7c42ca7357572f509cfd8d6298aa8e7a6955b4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty