Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydrazine acetate ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

H1112-25G H1112-5G
TCH1112-25GEA 322 USD InStock
TCH1112-25G TCH1112-5G
Hydrazine acetate ≥98.0% (by titrimetric analysis)
Hydrazine acetate
CAS Number: 13255-48-6
Molecular Formula: H4N2
Molecular Weight: 92.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 101


Formula: H₄N₂
Melting Pt: 101 °C
MDL Number: MFCD00013141
CAS Number: 13255-48-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=344ecd2ca992e1548f69f14f1925e158ddd08218&vid=344ecd2ca992e1548f69f14f1925e158ddd08218&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty