Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxythiophenol ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

H1177-25G H1177-5G
TCH1177-25GEA 269 USD InStock
TCH1177-25G TCH1177-5G
2-Hydroxythiophenol ≥97.0% (by titrimetric analysis)
2-Hydroxythiophenol
CAS Number: 1121-24-0
Molecular Formula: C6H6OS
Molecular Weight: 126.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 217
Melting point (°C): 6
Flash Point (°C): 110
Specific Gravity (20/20): 1.22

Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 2-Hydroxythiophenol, 2-Mercaptophenol


Formula: C₆H₆OS
Boiling Pt: 217 °C
Melting Pt: 6 °C
Density: 1.22 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00040447
CAS Number: 1121-24-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b09804e8eccd0371cb1740dbc2ffec8570aea403&vid=b09804e8eccd0371cb1740dbc2ffec8570aea403&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty