Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iodotrimethylsilane ≥95.0% stabilised

Supplier: Decon Laboratories Ltd

I0308-25G I0308-5G
TCI0308-25GEA 94.56 USD InStock
TCI0308-25G TCI0308-5G
Iodotrimethylsilane ≥95.0% stabilised
Iodotrimethylsilane
(stabilized with Aluminum)
CAS Number: 16029-98-4
MDL Number: MFCD00001028
Molecular Formula: C3H9ISi
Molecular Weight: 200.09
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): -31
Specific Gravity (20/20): 1.46

Product Categories: Synthetic Organic Chemistry, Halogenation, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Iodotrimethylsilane, Trimethyliodosilane


Formula: C₃H₉ISi
MW: 200.09 g/mol
Boiling Pt: 106
Density: 1.46 (20 °C)
Flash Pt: –31 °C
MDL Number: MFCD00001028
CAS Number: 16029-98-4
UN: 2924
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5795c365bcf0b20351542c3bd16860bfaa4c2bd&vid=e5795c365bcf0b20351542c3bd16860bfaa4c2bd&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty