Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

o-Tolyl isothiocyanate ≥99.0% (by GC)

Supplier: Decon Laboratories Ltd

I0458-25G I0458-5G
TCI0458-25GEA 259 USD InStock
TCI0458-25G TCI0458-5G
o-Tolyl isothiocyanate ≥99.0% (by GC)
o-Tolyl isothiocyanate
CAS Number: 614-69-7
MDL Number: MFCD00004802
Molecular Formula: C8H7NS
Molecular Weight: 149.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 239
Flash Point (°C): 107
Specific Gravity (20/20): 1.11

Other Product Names / Synonyms: Isothiocyanic Acid o-Tolyl Ester, 2-Methylphenyl Isothiocyanate


Formula: C₈H₇NS
Boiling Pt: 239 °C
Density: 1.11 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00004802
CAS Number: 614-69-7
UN: 2810
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f3e592bb414176cb516bce709f6ec75ab886e38&vid=4f3e592bb414176cb516bce709f6ec75ab886e38&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty