Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isobutylnitrite ≥95.0%

Supplier: Decon Laboratories Ltd

I0528-25ML I0528-500ML
TCI0528-25MLEA 25.47 USD InStock
TCI0528-25ML TCI0528-500ML
Isobutylnitrite ≥95.0%
IsobutylNitrite
CAS Number: 542-56-3
MDL Number: MFCD00002056
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): -21
Specific Gravity (20/20): 0.87
Storage Temperature: 0-10°C

Product Categories: Bioscience, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: 2-Methylpropyl Nitrite


Formula: C₄H₉NO₂
MW: 103.12
Boiling Pt: 67 °C
Density: 0.87 (20 °C)
Flash Pt: –21 °C
MDL Number: MFCD00002056
CAS Number: 542-56-3
UN: 2351
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0750675f7bc8689dec38dfe058e4f70c54f66e9b&vid=0750675f7bc8689dec38dfe058e4f70c54f66e9b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty