I0562-25G
I0562-5G
TCI0562-25GEA
301.1
USD
InStock
TCI0562-25G
TCI0562-5G
Ethyl isocyanoacetate ≥97.0%
Ethyl isocyanoacetate
CAS Number: 2999-46-4
MDL Number: MFCD00000007
Molecular Formula: C5H7NO2
Molecular Weight: 113.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 84
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Isonitriles (Multi-component Reaction)
Other Product Names / Synonyms: Isocyanoacetic Acid Ethyl Ester