Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indomethacin ≥98.0%

Supplier: Decon Laboratories Ltd

I0655-25G I0655-500G I0655-100G
TCI0655-25GEA 62.04 USD InStock
TCI0655-25G TCI0655-500G TCI0655-100G
Indomethacin ≥98.0%
Indomethacin
CAS Number: 53-86-1
MDL Number: MFCD00057095
Molecular Formula: C19H16ClNO4
Molecular Weight: 357.79
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 160

Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Reagents for Inflammation & Immunity Disorder Models, Regenerative Medicine Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingr
Other Product Names / Synonyms: 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-3-indoleacetic Acid


Formula: C₁₉H₁₆ClNO₄
MW: 357.80 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00057095
CAS Number: 53-86-1
UN: 2811
ADR: 6.1 I
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9472d51e6c32c287634b41faa91710fcd0b3079&vid=e9472d51e6c32c287634b41faa91710fcd0b3079&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty