Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Levamisole hydrochloride ≥99.0% (by titrimetric analysis) for biochemical research

Supplier: Decon Laboratories Ltd

L0231-1G L0231-5G
TCL0231-1GEA 47.75 USD InStock
TCL0231-1G TCL0231-5G
Levamisole hydrochloride ≥99.0% (by titrimetric analysis) for biochemical research
Levamisole hydrochloride
[for Biochemical Research]
CAS Number: 16595-80-5
MDL Number: MFCD00012675
Molecular Formula: C11H12N2S
Molecular Weight: 240.75
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 230
Specific rotation [a]20/D: -126 deg (C=1, H2O)

Product Categories: Bioscience, Alkaline Phosphatase Inhibitors (for Biochemical Research), Anthelmintic Ingredients
Other Product Names / Synonyms: (-)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole Hydrochloride, (-)-Tetramisole Hydrochloride


Formula: C₁₁H₁₂N₂S
Melting Pt: 230 °C
MDL Number: MFCD00012675
CAS Number: 16595-80-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5033cd3e010bc127d615ecfca53dea74ddd30c0d&vid=5033cd3e010bc127d615ecfca53dea74ddd30c0d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty