Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylpyrimidine ≥96.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

M0356-25G M0356-5G
TCM0356-25GEA 166.3 USD InStock
TCM0356-25G TCM0356-5G
4-Methylpyrimidine ≥96.0% (by titrimetric analysis)
4-Methylpyrimidine
CAS Number: 3438-46-8
MDL Number: MFCD00006115
Molecular Formula: C5H6N2
Molecular Weight: 94.12
Purity/Analysis Method: >96.0% (T)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 40
Specific Gravity (20/20): 1.02


Formula: C₅H₆N₂
Boiling Pt: 142 °C
Density: 1.02 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00006115
CAS Number: 3438-46-8
UN: 1993
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ebf137eecd5954cd0baf966fcb079a7ccac0ca4&vid=6ebf137eecd5954cd0baf966fcb079a7ccac0ca4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty