Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methylethylenediamine ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

M0518-25ML M0518-5ML
TCM0518-25MLEA 186 USD InStock
TCM0518-25ML TCM0518-5ML
N-Methylethylenediamine ≥97.0% (by titrimetric analysis)
N-Methylethylenediamine
CAS Number: 109-81-9
MDL Number: MFCD00008165
Molecular Formula: C3H10N2
Molecular Weight: 74.13
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 41
Specific Gravity (20/20): 0.85


Formula: C₃H₁₀N₂
Boiling Pt: 116 °C
Density: 0.85 (20 °C)
Flash Pt: 41 °C
MDL Number: MFCD00008165
CAS Number: 109-81-9
UN: 2734
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c8eabcdac345bd318d0cf7fdfd4938c913f37bc&vid=7c8eabcdac345bd318d0cf7fdfd4938c913f37bc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty