Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-N-trimethylsilylacetamide ≥95.0% (by GC)

Supplier: Decon Laboratories Ltd

M0536-10G M0536-25G
TCM0536-10GEA 93 USD InStock
TCM0536-10G TCM0536-25G
N-Methyl-N-trimethylsilylacetamide ≥95.0% (by GC)
N-Methyl-N-trimethylsilylacetamide
[Trimethylsilylating Agent]
CAS Number: 7449-74-3
MDL Number: MFCD00040758
Molecular Formula: C6H15NOSi
Molecular Weight: 145.28
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Flash Point (°C): 48
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: N-Methyl-N-TMS-acetamide


Formula: C₆H₁₅NOSi
Boiling Pt: 154 °C
Density: 0.91 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00040758
CAS Number: 7449-74-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=656c4adfd82712e4e18f6ec698359f23a54d252b&vid=656c4adfd82712e4e18f6ec698359f23a54d252b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty