Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxybenzylamine ≥97.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

M0870-250ML M0870-25ML
TCM0870-250MLEA 206 USD InStock
TCM0870-250ML TCM0870-25ML
4-Methoxybenzylamine ≥97.0% (by GC, titration analysis)
4-Methoxybenzylamine
CAS Number: 2393-23-9
MDL Number: MFCD00008122
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 236
Flash Point (°C): 110
Specific Gravity (20/20): 1.06


Formula: C₈H₁₁NO
Boiling Pt: 236 °C
Density: 1.06 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008122
CAS Number: 2393-23-9
UN: 2735
ADR: 8,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46b5dd34645167b4584890b1468d93d839336962&vid=46b5dd34645167b4584890b1468d93d839336962&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty