Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-2-pentanol ≥96.0% (by GC)

Supplier: Decon Laboratories Ltd

M1178-25ML M1178-5ML
TCM1178-25MLEA 140.2 USD InStock
TCM1178-25ML TCM1178-5ML
2-Methyl-2-pentanol ≥96.0% (by GC)
2-Methyl-2-pentanol
CAS Number: 590-36-3
MDL Number: MFCD00004485
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 30
Specific Gravity (20/20): 0.81

Other Product Names / Synonyms: Dimethylpropylcarbinol


Formula: C₆H₁₄O
Boiling Pt: 122 °C
Density: 0.81 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00004485
CAS Number: 590-36-3
UN: 2560
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9790aae0d4a18a3d09a008585a59fdf90e219e9&vid=b9790aae0d4a18a3d09a008585a59fdf90e219e9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty