Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Methylenanthic acid ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

M1265-25G M1265-5G
TCM1265-25GEA 105 USD InStock
TCM1265-25G TCM1265-5G
(±)-2-Methylenanthic acid ≥98.0% (by GC, titration analysis)
(±)-2-Methylenanthic acid
CAS Number: 1188-02-9
MDL Number: MFCD00142944
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (T,GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 109
Specific Gravity (20/20): 0.91

Other Product Names / Synonyms: 2-Methylenanthic Acid


Formula: C₈H₁₆O₂
Boiling Pt: 140 °C
Density: 0.91 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00142944
CAS Number: 1188-02-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6ec19ee5f96bd3f0762d4cc2063613612fb67bd&vid=d6ec19ee5f96bd3f0762d4cc2063613612fb67bd&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T,GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty