Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers) ≥99.0% (by GC)

Supplier: Decon Laboratories Ltd

M1422-25ML M1422-500ML
TCM1422-25MLEA 27.07 USD InStock
TCM1422-25ML TCM1422-500ML
4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers) ≥99.0% (by GC)
4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers)
CAS Number: 6864-37-5
MDL Number: MFCD00075535
Molecular Formula: C15H30N2
Molecular Weight: 238.42
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 0.95

Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: Bis(4-amino-3-methylcyclohexyl)methane, 4,4'-Diamino-3,3'-dimethyldicyclohexylmethane, 4,4'-Methylenebis(1-amino-2-methylcyclohexane)


Formula: C₁₅H₃₀N₂
Density: 0.95 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00075535
CAS Number: 6864-37-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7308beaa2641d1f5e581879d6eac09677ee7233a&vid=7308beaa2641d1f5e581879d6eac09677ee7233a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty