Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-2-Methylpiperazine ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

M1424-1G M1424-5G
TCM1424-1GEA 82.6 USD InStock
TCM1424-1G TCM1424-5G
(S)-(+)-2-Methylpiperazine ≥97.0% (by titrimetric analysis)
(S)-(+)-2-Methylpiperazine
CAS Number: 74879-18-8
MDL Number: MFCD00171452
Molecular Formula: C5H12N2
Molecular Weight: 100.17
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 91
Flash Point (°C): 65
Specific rotation [a]20/D: 7 deg (C=1, EtOH)


Formula: C₅H₁₂N₂
Melting Pt: 91 °C
Flash Pt: 65 °C
MDL Number: MFCD00171452
CAS Number: 74879-18-8
UN: 1325
ADR: 4.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42307b31dc612fe79ae889dfac6dc748034a43d0&vid=42307b31dc612fe79ae889dfac6dc748034a43d0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty