Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methylamino)-1-propanol ≥97.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

M1484-25G M1484-5G
TCM1484-25GEA 397 USD InStock
TCM1484-25G TCM1484-5G
3-(Methylamino)-1-propanol ≥97.0% (by GC, titration analysis)
3-(Methylamino)-1-propanol
CAS Number: 42055-15-2
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 169
Specific Gravity (20/20): 0.94


Formula: C₄H₁₁NO
Boiling Pt: 169 °C
Density: 0.94 (20 °C)
MDL Number: MFCD06797456
CAS Number: 42055-15-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcc3f52f4e231e18f00a65d6e553a3fbb01411a7&vid=bcc3f52f4e231e18f00a65d6e553a3fbb01411a7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty