Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methanedisulfonyl dichloride ≥96.0%

Supplier: Decon Laboratories Ltd

M1669-25G M1669-5G
TCM1669-25GEA 1008 USD InStock
TCM1669-25G TCM1669-5G
Methanedisulfonyl dichloride ≥96.0%
Methanedisulfonyl dichloride
CAS Number: 5799-68-8
Molecular Formula: CH2Cl2O4S2
Molecular Weight: 213.04
Purity/Analysis Method: >96.0% (T)
Form: Clear Liquid
Boiling point (°C): 138
Specific Gravity (20/20): 1.83

Other Product Names / Synonyms: Methylenedisulfonyl Dichloride, Methanedisulfonyl Chloride


Formula: CH₂Cl₂O₄S₂
MW: 213.04 g/mol
Boiling Pt: 138 °C
Density: 1.83 (20 °C)
MDL Number: MFCD00176854
CAS Number: 5799-68-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10a16c6987bf3105f6077548ee45ad30d0f27103&vid=10a16c6987bf3105f6077548ee45ad30d0f27103&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty