Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl tridecafluoroheptanoate ≥96.0% (by GC)

Supplier: Decon Laboratories Ltd

M1914-25G M1914-5G
TCM1914-25GEA 183 USD InStock
TCM1914-25G TCM1914-5G
Methyl tridecafluoroheptanoate ≥96.0% (by GC)
Methyl tridecafluoroheptanoate
CAS Number: 14312-89-1
MDL Number: MFCD00153212
Molecular Formula: C8H3F13O2
Molecular Weight: 378.09
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Specific Gravity (20/20): 1.66

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds
Other Product Names / Synonyms: Methyl Perfluoroheptanoate, Tridecafluoroheptanoic Acid Methyl Ester, Perfluoroheptanoic Acid Methyl Ester


Formula: C₈H₃F₁₃O₂
Boiling Pt: 141 °C
Density: 1.66 (20 °C)
MDL Number: MFCD00153212
CAS Number: 14312-89-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23f423c97f634919123e977197cf3682d47b8168&vid=23f423c97f634919123e977197cf3682d47b8168&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty