Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Meloxicam ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd

M1959-25G M1959-5G
TCM1959-25GEA 180 USD InStock
TCM1959-25G TCM1959-5G
Meloxicam ≥98.0% (by HPLC)
Meloxicam
CAS Number: 71125-38-7
MDL Number: MFCD00868752
Molecular Formula: C14H13N3O4S2
Molecular Weight: 351.40
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 255
Storage Temperature: 0-10°C

Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide


Formula: C₁₄H₁₃N₃O₄S₂
Melting Pt: 255 °C
MDL Number: MFCD00868752
CAS Number: 71125-38-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aaf5808643de2f91a5ba7f4f95da6e6a1d083e20&vid=aaf5808643de2f91a5ba7f4f95da6e6a1d083e20&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty