Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(1-Naphthyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Supplier: Decon Laboratories Ltd

N0071-25G N0071-5G
TCN0071-25GEA 170 USD InStock
TCN0071-25G TCN0071-5G
1-(1-Naphthyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
1-(1-Naphthyl)-2-thiourea
CAS Number: 86-88-4
MDL Number: MFCD00041824
Molecular Formula: C11H10N2S
Molecular Weight: 202.28
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 191


Formula: C₁₀H₇NHC(S)NH₂
Melting Pt: 191 °C
MDL Number: MFCD00041824
CAS Number: 86-88-4
UN: 1651
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65719c5f9c143783b29e9cd52d953c47e65eaa08&vid=65719c5f9c143783b29e9cd52d953c47e65eaa08&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty