Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H,2H-Perfluoro-1-hexene ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

N0601-25G N0601-5G
TCN0601-25GEA 76 USD InStock
TCN0601-25G TCN0601-5G
1H,1H,2H-Perfluoro-1-hexene ≥98.0% (by GC)
1H,1H,2H-Perfluoro-1-hexene
CAS Number: 19430-93-4
MDL Number: MFCD00042338
Molecular Formula: C6H3F9
Molecular Weight: 246.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): -17
Specific Gravity (20/20): 1.45

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: (Perfluorobutyl)ethylene


Formula: C₆H₃F₉
Boiling Pt: 60 °C
Density: 1.45 (20 °C)
Flash Pt: –17 °C
MDL Number: MFCD00042338
CAS Number: 19430-93-4
UN: 1993
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=661b242bd50fdc0394aa50c89e82e4a2e70a986c&vid=661b242bd50fdc0394aa50c89e82e4a2e70a986c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty