Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nonafluorovaleric acid ≥98.0%

Supplier: Decon Laboratories Ltd

N0605-25G N0605-5G
TCN0605-25GEA 310.48 USD InStock
TCN0605-25G TCN0605-5G
Nonafluorovaleric acid ≥98.0%
Nonafluorovaleric acid
CAS Number: 2706-90-3
MDL Number: MFCD00040211
Molecular Formula: C5HF9O2
Molecular Weight: 264.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 70
Specific Gravity (20/20): 1.71

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Nonafluoropentanoic Acid, Perfluoropentanoic Acid, Perfluorovaleric Acid


Formula: C₅HF₉O₂
MW: 264.05 g/mol
Boiling Pt: 70 °C
Density: 1.71 (20 °C)
MDL Number: MFCD00040211
CAS Number: 2706-90-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=692f5faddc11b4f8d86ee5e4efc3d170b5aa2e96&vid=692f5faddc11b4f8d86ee5e4efc3d170b5aa2e96&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty