Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palmitoyl chloride ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

P0009-25ML P0009-500ML
TCP0009-25MLEA 31.48 USD InStock
TCP0009-25ML TCP0009-500ML
Palmitoyl chloride ≥97.0% (by titrimetric analysis)
Palmitoyl chloride
CAS Number: 112-67-4
MDL Number: MFCD00000742
Molecular Formula: C16H31ClO
Molecular Weight: 274.87
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 166
Freezing point (°C): 10
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C


Formula: C₁₆H₃₁ClO
MDL Number: MFCD00000742
CAS Number: 112-67-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=449311d9c690701e71e98abe41e00535c7c35baf&vid=449311d9c690701e71e98abe41e00535c7c35baf&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty