P0043-25G
P0043-5G
TCP0043-25GEA
218
USD
InStock
TCP0043-25G
TCP0043-5G
Pentamethylbenzene ≥99.0%
Pentamethylbenzene
CAS Number: 700-12-9
MDL Number: MFCD00008522
Molecular Formula: C11H16
Molecular Weight: 148.25
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 231
Melting point (°C): 53
Flash Point (°C): 91