P0566-25G
P0566-5G
TCP0566-25GEA
211
USD
InStock
TCP0566-25G
TCP0566-5G
Pentafluoropropionic anhydride ≥95.0%
Pentafluoropropionic anhydride
CAS Number: 356-42-3
MDL Number: MFCD00000429
Molecular Formula: C6F10O3
Molecular Weight: 310.05
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Specific Gravity (20/20): 1.59
Product Categories: Synthetic Organic Chemistry, Other Fluorinated Group Introducing Agents, Analytical Science, Acylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: Decafluoropropionic Anhydride, Perfluoropropionic Anhydride