P0856-25G
P0856-5G
TCP0856-25GEA
73.88
USD
InStock
TCP0856-25G
TCP0856-5G
Perfluorotoluene ≥98.0%
Perfluorotoluene
CAS Number: 434-64-0
MDL Number: MFCD00000375
Molecular Formula: C7F8
Molecular Weight: 236.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 20
Specific Gravity (20/20): 1.68
Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluorotoluene