Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Biphenylcarbonyl chloride ≥98.0%

Supplier: Decon Laboratories Ltd

P1079-25G P1079-5G
TCP1079-25GEA 145 USD InStock
TCP1079-25G TCP1079-5G
4-Biphenylcarbonyl chloride ≥98.0%
4-Biphenylcarbonyl chloride
CAS Number: 14002-51-8
MDL Number: MFCD00000692
Molecular Formula: C13H9ClO
Molecular Weight: 216.66
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 160
Melting point (°C): 113

Product Categories: Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: 4-Biphenylcarbonyl Chloride


Formula: C₁₃H₉ClO
MW: 216.67 g/mol
Boiling Pt: 160 °C
Melting Pt: 113 °C
MDL Number: MFCD00000692
CAS Number: 14002-51-8
UN: 3261
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=874d4967bc236d23f4b7886cd9df77055fb967eb&vid=874d4967bc236d23f4b7886cd9df77055fb967eb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty