Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H,2H-Perfluoro-1-octene ≥96.0% (by GC)

Supplier: Decon Laboratories Ltd

P1102-25G P1102-100G
TCP1102-10GEA 26.64 USD InStock
TCP1102-10G TCP1102-25G TCP1102-100G
1H,1H,2H-Perfluoro-1-octene ≥96.0% (by GC)
1H,1H,2H-Perfluoro-1-octene
CAS Number: 25291-17-2
MDL Number: MFCD00039249
Molecular Formula: C8H3F13
Molecular Weight: 346.09
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 103
Specific Gravity (20/20): 1.58

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds
Other Product Names / Synonyms: (Tridecafluorohexyl)ethylene


Formula: C₈H₃F₁₃
Boiling Pt: 103 °C
Density: 1.58 (20 °C)
MDL Number: MFCD00039249
CAS Number: 25291-17-2
UN: 1993
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e28ee71d4a5ba1df4da0ded3e0bd88798ae5100&vid=8e28ee71d4a5ba1df4da0ded3e0bd88798ae5100&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty