Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Phenylthio)trimethylsilane ≥95.0%

Supplier: Decon Laboratories Ltd

P1378-25G P1378-5G
TCP1378-25GEA 348 USD InStock
TCP1378-25G TCP1378-5G
(Phenylthio)trimethylsilane ≥95.0%
(Phenylthio)trimethylsilane
CAS Number: 4551-15-9
MDL Number: MFCD00008272
Molecular Formula: C9H14SSi
Molecular Weight: 182.36
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 65
Flash Point (°C): 33
Specific Gravity (20/20): 0.97

Product Categories: Synthetic Organic Chemistry, Sulfenylation, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Phenyl Trimethylsilyl Sulfide, Trimethyl(phenylthio)silane, S-(Trimethylsilyl)benzenethiol


Formula: C₉H₁₄SSi
MW: 182.36 g/mol
Boiling Pt: 65 °C
Density: 0.97 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00008272
CAS Number: 4551-15-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4be418c0d2e0b6063f75bcfe5e8f5fe066876ec&vid=c4be418c0d2e0b6063f75bcfe5e8f5fe066876ec&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty