Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pivalic anhydride ≥98.0%

Supplier: Decon Laboratories Ltd

P1414-250ML P1414-25ML
TCP1414-250MLEA 281 USD InStock
TCP1414-250ML TCP1414-25ML
Pivalic anhydride ≥98.0%
Pivalic anhydride
CAS Number: 1538-75-6
MDL Number: MFCD00008842
Molecular Formula: C10H18O3
Molecular Weight: 186.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 57
Specific Gravity (20/20): 0.92

Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: 2,2-Dimethylpropionic Anhydride, Trimethylacetic Anhydride


Formula: C₁₀H₁₈O₃
MW: 186.25 g/mol
Boiling Pt: 193
Density: 0.92 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00008842
CAS Number: 1538-75-6
UN: 3265
ADR: 8 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f90ab3baa6f4edc490da29b6673f25d2fa54d2e&vid=3f90ab3baa6f4edc490da29b6673f25d2fa54d2e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty