P1665-25G
P1665-5G
TCP1665-25GEA
84
USD
InStock
TCP1665-25G
TCP1665-5G
2-Phenylbutyryl chloride ≥97.0% (by GC, titration analysis)
2-Phenylbutyryl chloride
CAS Number: 36854-57-6
MDL Number: MFCD00018811
Molecular Formula: C10H11ClO
Molecular Weight: 182.65
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 125
Flash Point (°C): 98
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: 2-Ethyl-2-phenylacetyl Chloride