Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenethyl isocyanate ≥98.0%

Supplier: Decon Laboratories Ltd

P1677-25G P1677-5G
TCP1677-25GEA 59.24 USD InStock
TCP1677-25G TCP1677-5G
Phenethyl isocyanate ≥98.0%
Phenethyl isocyanate
CAS Number: 1943-82-4
MDL Number: MFCD00037044
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 210
Flash Point (°C): 100
Specific Gravity (20/20): 1.06

Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: 2-Phenylethyl Isocyanate, (2-Isocyanatoethyl)benzene, Isocyanic Acid Phenethyl Ester, Isocyanic Acid 2-Phenylethyl Ester


Formula: C₉H₉NO
Boiling Pt: 210 °C
Density: 1.06 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00037044
CAS Number: 1943-82-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8e8549e2f95ab4c42c14513f87d7aa542832689&vid=c8e8549e2f95ab4c42c14513f87d7aa542832689&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty