P1686-25G
P1686-500G
TCP1686-25GEA
32.56
USD
InStock
TCP1686-25G
TCP1686-500G
Phenylbutazone ≥98.0% (by HPLC, titration analysis)
Phenylbutazone
CAS Number: 50-33-9
MDL Number: MFCD00005500
Molecular Formula: C19H20N2O2
Molecular Weight: 308.38
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 106
Other Product Names / Synonyms: 4-Butyl-1,2-diphenylpyrazolidine-3,5-dione