Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Quinolinesulphonyl chloride ≥98.0%

Supplier: Decon Laboratories Ltd

Q0016-25G Q0016-5G
TCQ0016-25GEA 239.6 USD InStock
TCQ0016-25G TCQ0016-5G
8-Quinolinesulphonyl chloride ≥98.0%
8-Quinolinesulphonyl chloride
CAS Number: 18704-37-5
MDL Number: MFCD00006808
Molecular Formula: C9H6ClNO2S
Molecular Weight: 227.66
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 306
Melting point (°C): 128


Formula: C₉H₆ClNO₂S
MW: 227.67 g/mol
Boiling Pt: 306 °C
Melting Pt: 128 °C
MDL Number: MFCD00006808
CAS Number: 18704-37-5
UN: 3261
ADR: 8 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cac435f806dc4b5697fe7a18d3b3d1b16fd555a&vid=2cac435f806dc4b5697fe7a18d3b3d1b16fd555a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty