Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Sulfophthalic Acid ∼50% in water (contains 3-Sulfophthalic Acid)

Supplier: Decon Laboratories Ltd

S0136-25ML S0136-500ML
TCS0136-25MLEA 20 USD InStock
TCS0136-25ML TCS0136-500ML
4-Sulfophthalic Acid ∼50% in water (contains 3-Sulfophthalic Acid)
4-Sulfophthalic Acid
CAS Number: 89-08-7
MDL Number: MFCD00007494
Molecular Formula: C8H6O7S
Molecular Weight: 246.19
Form: Clear Liquid
Color: Pale Yellow
Specific Gravity (20/20): 1.29


Formula: C₈H₆O₇S
Density: 1.29 (20 °C)
MDL Number: MFCD00007494
CAS Number: 89-08-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971998128908e7c469bb97b073af0155014acdb7&vid=971998128908e7c469bb97b073af0155014acdb7&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty